Methyl oxalyl chloride 96%

Supplier: Apollo Scientific

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

OR924197-100G OR924197-25G OR924197-5G
APOSOR924197-100GEA 459 EUR
APOSOR924197-100G APOSOR924197-25G APOSOR924197-5G
Methyl oxalyl chloride 96%
Methyl oxalyl chloride
Formula: C₃H₃ClO₃
MW: 122,51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000705
CAS Number: 5781-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR