5-Bromo-1H-indazole 97%
Supplier: Apollo Scientific
|
Danger
|
OR9243-1G
OR9243-25G
OR9243-5G
APOSOR9243-1GEA
14.7
EUR
APOSOR9243-1G
APOSOR9243-25G
APOSOR9243-5G
5-Bromo-1H-indazole 97%
5-Bromo-1H-indazole
Formula:
C₇H₅BrN₂ MW: 197,03 g/mol Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00839493 CAS Number: 53857-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...