Glutaric anhydride 95%

Supplier: Apollo Scientific

Synonyms: Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione

OR924369-100G OR924369-500G
APOSOR924369-100GEA 55.3 EUR
APOSOR924369-100G APOSOR924369-500G
Glutaric anhydride 95%
Glutaric anhydride
Formula: C₅H₆O₃
MW: 114,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006679
CAS Number: 108-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR