(1R)-Camphor oxime 95%
Supplier: Apollo Scientific
OR924411-1G
OR924411-5G
APOSOR924411-1GEA
132
EUR
APOSOR924411-1G
APOSOR924411-5G
(1R)-Camphor oxime 95%
(1R)-Camphor oxime
Formula:
C₁₀H₁₇NO MW: 167,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046298 CAS Number: 2792-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...