tert-Amyl ethyl ether 95%

Supplier: Apollo Scientific

OR924485-1G OR924485-25G OR924485-5G
APOSOR924485-1GEA 30.1 EUR
APOSOR924485-1G APOSOR924485-25G APOSOR924485-5G
tert-Amyl ethyl ether 95%
Formula: C₇H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD01727266
CAS Number: 919-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR