5-Ketohexanenitrile 95%
Supplier: Apollo Scientific
Synonyms:
4-Acetylbutyronitrile, 5-Oxohexanenitrile
OR924589-1G
OR924589-25G
OR924589-5G
APOSOR924589-1GEA
37.7
EUR
APOSOR924589-1G
APOSOR924589-25G
APOSOR924589-5G
5-Ketohexanenitrile 95%
5-Ketohexanenitrile
Formula:
C₆H₉NO MW: 111.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010671 CAS Number: 10412-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...