Isopropyl chloroacetate 95%
Supplier: Apollo Scientific
OR924675-100G
OR924675-500G
APOSOR924675-100GEA
145
EUR
APOSOR924675-100G
APOSOR924675-500G
Isopropyl chloroacetate 95%
Isopropyl chloroacetate
Formula:
C₅H₉ClO₂ MW: 136,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00040410 CAS Number: 105-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...