Ethyl-4-(Chloromethyl)benzoate 95%

Supplier: Apollo Scientific

Synonyms: 4-(Chloromethyl)benzoic Acid Ethyl Ester, Ethyl 4-(chloromethyl)benzoate

OR924738-1G OR924738-25G OR924738-5G
APOSOR924738-1GEA 118 EUR
APOSOR924738-1G APOSOR924738-25G APOSOR924738-5G
Ethyl-4-(Chloromethyl)benzoate 95%
Ethyl-4-(Chloromethyl)benzoate
Formula: C₁₀H₁₁ClO₂
MW: 198,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04337336
CAS Number: 1201-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR