2-Phenoxybenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
OR9248-1G
APOSOR9248-1GEA
248
EUR
APOSOR9248-1G
2-Phenoxybenzyl bromide
2-Phenoxybenzyl bromide
Formula:
C₁₃H₁₁BrO MW: 263,134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01320513 CAS Number: 82657-72-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...