3-Methyl-4-pyrrolidin-1-ylbenzaldehyde 95%

Supplier: Apollo Scientific

OR924984-100MG OR924984-1G OR924984-250MG
APOSOR924984-100MGEA 190 EUR
APOSOR924984-100MG APOSOR924984-1G APOSOR924984-250MG
3-Methyl-4-pyrrolidin-1-ylbenzaldehyde 95%
3-Methyl-4-pyrrolidin-1-ylbenzaldehyde
Formula: C₁₂H₁₅NO
MW: 189,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03063768
CAS Number: 461033-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR