Ethyl benzoate 95%

Supplier: Apollo Scientific

Synonyms: Benzoic acid ethyl ester

OR925081-100G OR925081-1KG OR925081-500G
APOSOR925081-100GEA 62.5 EUR
APOSOR925081-100G APOSOR925081-1KG APOSOR925081-500G
Ethyl benzoate 95%
Ethyl benzoate
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR