4-Pentylpyridine 95%
Supplier: Apollo Scientific
Synonyms:
4-Amylpyridine, 4-Pentylpyridine
OR925136-1G
OR925136-5G
APOSOR925136-1GEA
127
EUR
APOSOR925136-1G
APOSOR925136-5G
4-Pentylpyridine 95%
4-Pentylpyridine
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093422 CAS Number: 2961-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...