2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one 97%

Supplier: Apollo Scientific

Synonyms: 2,2-Dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one

OR925185-1G OR925185-250MG
APOSOR925185-1GEA 270 EUR
APOSOR925185-1G APOSOR925185-250MG
2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one 97%
2,2-Dimethyl-2H-1,4-benzoxazin-3(4H)-one
Formula: C₁₀H₁₁NO₂
MW: 177,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11868710
CAS Number: 10514-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR