1-Benzyl-1,2,3,6-tetrahydropyridine 95%

Supplier: Apollo Scientific

OR925565-25G OR925565-5G
APOSOR925565-25GEA 121 EUR
APOSOR925565-25G APOSOR925565-5G
1-Benzyl-1,2,3,6-tetrahydropyridine 95%
1-Benzyl-1,2,3,6-tetrahydropyridine
Formula: C₁₂H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD11501660
CAS Number: 40240-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR