3-Iodo-1H-pyrrolo[2,3-c]pyridine ≥98%

Supplier: Apollo Scientific

OR925581-1G OR925581-250MG
APOSOR925581-1GEA 72.7 EUR
APOSOR925581-1G APOSOR925581-250MG
3-Iodo-1H-pyrrolo[2,3-c]pyridine ≥98%
3-Iodo-1H-pyrrolo[2,3-c]pyridine
Formula: C₇H₅IN₂
Storage Temperature: Ambient
MDL Number: MFCD10699183
CAS Number: 956003-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR