6-Chloro-N,N,2-trimethylpyrimidin-4-amine 95%

Supplier: Apollo Scientific

Synonyms: 4-(N,N-Dimethylamino)-6-chloro-2-methylpyrimidine

OR925862-1G OR925862-5G
APOSOR925862-1GEA 58 EUR
APOSOR925862-1G APOSOR925862-5G
6-Chloro-N,N,2-trimethylpyrimidin-4-amine 95%
6-Chloro-N,N,2-trimethylpyrimidin-4-amine
Formula: C₇H₁₀ClN₃
Storage Temperature: Ambient
MDL Number: MFCD07782030
CAS Number: 58514-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR