3-(Methylthio)propyl acetate 95%
Supplier: Apollo Scientific
Synonyms:
Acetic Acid 3-(Methylthio)propyl Ester
OR925951-25G
APOSOR925951-25GEA
59.6
EUR
APOSOR925951-25G
3-(Methylthio)propyl acetate 95%
3-(Methylthio)propyl acetate
Formula:
C₆H₁₂O₂S MW: 148,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00053113 CAS Number: 16630-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...