6-Ethoxy-5-indanecarbaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
6-Ethoxy-2,3-dihydro-1H-indene-5-carbaldehyde
OR925983-1G
OR925983-5G
APOSOR925983-1GEA
256
EUR
APOSOR925983-1G
APOSOR925983-5G
6-Ethoxy-5-indanecarbaldehyde 95%
6-Ethoxy-5-indanecarbaldehyde
Formula:
C₁₂H₁₄O₂ MW: 190.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06247351 CAS Number: 876717-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...