5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid 95%
Supplier: Apollo Scientific
Synonyms:
5,6,7,8-Tetrahydro-2-naphthoic acid
OR926035-25G
OR926035-5G
APOSOR926035-25GEA
542
EUR
APOSOR926035-25G
APOSOR926035-5G
5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid 95%
5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861899 CAS Number: 1131-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...