Ethyl-2,4-dimethylpyrimidine-5-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 2,4-dimethylpyrimidine-5-carboxylate

OR926136-10G OR926136-1G OR926136-5G
APOSOR926136-10GEA 806 EUR
APOSOR926136-10G APOSOR926136-1G APOSOR926136-5G
Ethyl-2,4-dimethylpyrimidine-5-carboxylate 95%
Ethyl-2,4-dimethylpyrimidine-5-carboxylate
Formula: C₉H₁₂N₂O₂
MW: 180,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234561
CAS Number: 2226-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR