Ethyl-3-methylisoxazole-4-carboxylate 95%
Supplier: Apollo Scientific
OR926281-1G
OR926281-5G
APOSOR926281-1GEA
226
EUR
APOSOR926281-1G
APOSOR926281-5G
Ethyl-3-methylisoxazole-4-carboxylate 95%
Ethyl-3-methylisoxazole-4-carboxylate
Formula:
C₇H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD01075736 CAS Number: 20328-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...