O-4-Chlorobenzoylhydroxylamine 95%

Supplier: Apollo Scientific

OR926311-1G OR926311-5G
APOSOR926311-1GEA 190 EUR
APOSOR926311-1G APOSOR926311-5G
O-4-Chlorobenzoylhydroxylamine 95%
O-4-Chlorobenzoylhydroxylamine
Formula: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD22370138
CAS Number: 872851-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR