1-(3-Cyanopropyl)pyrrolidine 95%

Supplier: Apollo Scientific

Synonyms: 4-Pyrrolidinobutyronitrile, 1-Pyrrolidinebutanenitrile, 1-Pyrrolidinebutyronitrile

OR926503-100G OR926503-25G
APOSOR926503-100GEA 319 EUR
APOSOR926503-100G APOSOR926503-25G
1-(3-Cyanopropyl)pyrrolidine 95%
1-(3-Cyanopropyl)pyrrolidine
Formula: C₈H₁₄N₂
MW: 138,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003183
CAS Number: 35543-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR