2-Methylindazole-6-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methyl-2H-indazole-6-carboxylic acid

OR926826-1G OR926826-250MG
APOSOR926826-1GEA 128 EUR
APOSOR926826-1G APOSOR926826-250MG
2-Methylindazole-6-carboxylic acid 95%
2-Methylindazole-6-carboxylic acid
Formula: C₉H₈N₂O₂
MW: 176,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15071449
CAS Number: 1031417-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR