Vinyl crotonate 95%
Supplier: Apollo Scientific
Synonyms:
Crotonic Acid Vinyl Ester
OR927024-100G
OR927024-25G
OR927024-500G
APOSOR927024-100GEA
270
EUR
APOSOR927024-100G
APOSOR927024-25G
APOSOR927024-500G
Vinyl crotonate 95%
Vinyl crotonate
Formula:
C₆H₈O₂ MW: 112,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009288 CAS Number: 14861-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...