4-Nitrophthalide 97%

Supplier: Apollo Scientific

OR927035-1G OR927035-5G
APOSOR927035-1GEA 101 EUR
APOSOR927035-1G APOSOR927035-5G
4-Nitrophthalide 97%
4-Nitrophthalide
Formula: C₈H₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD03069001
CAS Number: 65399-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR