2-(4-Ethylphenyl)piperazine 95%
Supplier: Apollo Scientific
OR927066-1G
OR927066-250MG
APOSOR927066-1GEA
517
EUR
APOSOR927066-1G
APOSOR927066-250MG
2-(4-Ethylphenyl)piperazine 95%
2-(4-Ethylphenyl)piperazine
Formula:
C₁₂H₁₈N₂ Storage Temperature: Ambient |
CAS Number:
910444-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...