3-(2H-1,2,3-Triazol-2-yl)aniline 95%
Supplier: Apollo Scientific
OR927084-1G
OR927084-250MG
APOSOR927084-1GEA
488
EUR
APOSOR927084-1G
APOSOR927084-250MG
3-(2H-1,2,3-Triazol-2-yl)aniline 95%
3-(2H-1,2,3-Triazol-2-yl)aniline
Formula:
C₈H₈N₄ Storage Temperature: Ambient |
CAS Number:
626248-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...