4-(Dimethylamino)phenethyl alcohol 98%

Supplier: Apollo Scientific

Synonyms: 2-[4-(dimethylamino)phenyl]ethanol, 2-(4-(Dimethylamino)phenyl)ethanol, Benzeneethanol, 4-(dimethylamino)-, p-(Dimethylamino)phenethyl alcohol

OR927173-1G OR927173-5G
APOSOR927173-1GEA 154 EUR
APOSOR927173-1G APOSOR927173-5G
4-(Dimethylamino)phenethyl alcohol 98%
4-(Dimethylamino)phenethyl alcohol
Formula: C₁₀H₁₅NO
MW: 165,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009711
CAS Number: 50438-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR