(4-Methylfurazan-3-yl)acetic acid 95%
Supplier: Apollo Scientific
OR927310-1G
OR927310-250MG
APOSOR927310-1GEA
864
EUR
APOSOR927310-1G
APOSOR927310-250MG
(4-Methylfurazan-3-yl)acetic acid 95%
(4-Methylfurazan-3-yl)acetic acid
Formula:
C₅H₆N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD06010348 CAS Number: 15323-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...