3,4,5-Trimethoxyphenol 95%

Supplier: Apollo Scientific

Synonyms: Antiarol

OR927502-1G OR927502-25G OR927502-5G
APOSOR927502-1GEA 132 EUR
APOSOR927502-1G APOSOR927502-25G APOSOR927502-5G
3,4,5-Trimethoxyphenol 95%
3,4,5-Trimethoxyphenol
Formula: C₉H₁₂O₄
MW: 184,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008389
CAS Number: 642-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR