5-Nitro-m-toluonitrile 95%

Supplier: Apollo Scientific

Synonyms: 3-Methyl-5-nitrobenzonitrile

OR927557-1G
APOSOR927557-1GEA 77 EUR
APOSOR927557-1G
5-Nitro-m-toluonitrile 95%
5-Nitro-m-toluonitrile
Formula: C₈H₆N₂O₂
MW: 162,150 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17170423
CAS Number: 124289-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR