Mephenesin 95%
Supplier: Apollo Scientific
Synonyms:
3-(2-Methylphenoxy)-1,2-propanediol, 3-(o-Tolyloxy)propane-1,2-diol
OR927694-100G
OR927694-25G
APOSOR927694-100GEA
408
EUR
APOSOR927694-100G
APOSOR927694-25G
Mephenesin 95%
Mephenesin
Formula:
C₁₀H₁₄O₃ MW: 182,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004718 CAS Number: 59-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...