1-Methyl-2-phenylpiperazine dihydrochloride 95%

Supplier: Apollo Scientific

Synonyms: 1-Methyl-2-phenylpiperazine

OR927774-1G OR927774-250MG
APOSOR927774-1GEA 648 EUR
APOSOR927774-1G APOSOR927774-250MG
1-Methyl-2-phenylpiperazine dihydrochloride 95%
1-Methyl-2-phenylpiperazine dihydrochloride
Formula: C₁₁H₁₈Cl₂N₂
MW: 249,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801077
CAS Number: 5271-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR