2,7-Dimethylquinoline 95%

Supplier: Apollo Scientific
Warning

OR928151-1G OR928151-5G
APOSOR928151-1GEA 430 EUR
APOSOR928151-1G APOSOR928151-5G
2,7-Dimethylquinoline 95%
2,7-Dimethylquinoline
Formula: C₁₁H₁₁N
MW: 157,22 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006763
CAS Number: 93-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR