3-Hydroxy-5-methylbenzonitrile 98%

Supplier: Apollo Scientific

OR928299-1G OR928299-250MG
APOSOR928299-1GEA 531 EUR
APOSOR928299-1G APOSOR928299-250MG
3-Hydroxy-5-methylbenzonitrile 98%
3-Hydroxy-5-methylbenzonitrile
Formula: C8H7NO
MW: 133,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999137
CAS Number: 95658-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR