2-Chloro-5-methylthiazole 97%

Supplier: Apollo Scientific

OR928350-10G OR928350-1G OR928350-5G
APOSOR928350-10GEA 561 EUR
APOSOR928350-10G APOSOR928350-1G APOSOR928350-5G
2-Chloro-5-methylthiazole 97%
2-Chloro-5-methylthiazole
Formula: C₄H₄ClNS
MW: 133,6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03265390
CAS Number: 33342-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR