2-Ethyl-5-nitroaniline 98%
Supplier: Apollo Scientific
Synonyms:
2-Ethyl-5-nitrobenzenamine
OR928420-100G
OR928420-25G
OR928420-5G
APOSOR928420-100GEA
452
EUR
APOSOR928420-100G
APOSOR928420-25G
APOSOR928420-5G
2-Ethyl-5-nitroaniline 98%
2-Ethyl-5-nitroaniline
Formula:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00034056 CAS Number: 20191-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...