1H-Pyrazol-1-ylacetonitrile 95%

Supplier: Apollo Scientific

OR928801-1G OR928801-250MG
APOSOR928801-1GEA 502 EUR
APOSOR928801-1G APOSOR928801-250MG
1H-Pyrazol-1-ylacetonitrile 95%
1H-Pyrazol-1-ylacetonitrile
Formula: C₅H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD04971123
CAS Number: 113336-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR