Ethyl-2-chloropyridine-3-acetate 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-(2-chloropyridin-3-yl)acetate

OR928994-1G OR928994-250MG
APOSOR928994-1GEA 147 EUR
APOSOR928994-1G APOSOR928994-250MG
Ethyl-2-chloropyridine-3-acetate 95%
Ethyl-2-chloropyridine-3-acetate
Formula: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17214401
CAS Number: 164464-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR