Ethyl-6-aminopicolinate 95%

Supplier: Apollo Scientific

Synonyms: 6-Aminopyridine-2-carboxylic acid ethyl ester, Ethyl 6-Aminopicolinate, Ethyl 6-Aminopyridine-2-carboxylate, 6-Aminopicolinic acid ethyl ester, Ethyl 6-aminopicolinate

OR929091-1G OR929091-250MG
APOSOR929091-1GEA 59.7 EUR
APOSOR929091-1G APOSOR929091-250MG
Ethyl-6-aminopicolinate 95%
Ethyl-6-aminopicolinate
Formula: C₈H₁₀N₂O₂
MW: 166,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01009842
CAS Number: 69142-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR