4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane 95%
Supplier: Apollo Scientific
OR929253-1G
OR929253-250MG
APOSOR929253-1GEA
778
EUR
APOSOR929253-1G
APOSOR929253-250MG
4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane 95%
4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane
Formula:
C₆H₁₁BrO₂ MW: 195,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14584455 CAS Number: 36236-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...