3-Chloro-4-ethoxybenzaldehyde 95%

Supplier: Apollo Scientific

OR929320-1G OR929320-5G
APOSOR929320-1GEA 82.9 EUR
APOSOR929320-1G APOSOR929320-5G
3-Chloro-4-ethoxybenzaldehyde 95%
3-Chloro-4-ethoxybenzaldehyde
Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD06246171
CAS Number: 99585-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR