5-(m-Tolyl)pyrimidin-2-amine 95%

Supplier: Apollo Scientific

OR929358-1G OR929358-250MG
APOSOR929358-1GEA 719 EUR
APOSOR929358-1G APOSOR929358-250MG
5-(m-Tolyl)pyrimidin-2-amine 95%
5-(m-Tolyl)pyrimidin-2-amine
Formula: C₁₁H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD05864793
CAS Number: 914349-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR