5-Azidopentan-1-amine 95%
Supplier: Apollo Scientific
OR929551-1G
OR929551-250MG
APOSOR929551-1GEA
807
EUR
APOSOR929551-1G
APOSOR929551-250MG
5-Azidopentan-1-amine 95%
5-Azidopentan-1-amine
Formula:
C₅H₁₂N₄ Storage Temperature: Ambient |
MDL Number:
MFCD20275207 CAS Number: 148759-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...