2-Chloro-3-hydroxy-4-methoxybenzaldehyde 95%
Supplier: Apollo Scientific
Synonyms:
2-Chloro-3-hydroxy-p-anisaldehyde
OR929791-1G
OR929791-5G
APOSOR929791-1GEA
53.9
EUR
APOSOR929791-1G
APOSOR929791-5G
2-Chloro-3-hydroxy-4-methoxybenzaldehyde 95%
2-Chloro-3-hydroxy-4-methoxybenzaldehyde
Formula:
C₈H₇ClO₃ MW: 186,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425852 CAS Number: 37687-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...