N-Methyl-1-quinolin-8-ylmethanamine 95%

Supplier: Apollo Scientific

Synonyms: N-Methyl-1-(quinolin-8-yl)methanamine

OR929886-1G OR929886-250MG
APOSOR929886-1GEA 383 EUR
APOSOR929886-1G APOSOR929886-250MG
N-Methyl-1-quinolin-8-ylmethanamine 95%
N-Methyl-1-quinolin-8-ylmethanamine
Formula: C₁₁H₁₂N₂
MW: 172,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738895
CAS Number: 60843-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR