4,5-Dimethoxy-1-benzocyclobutenecarbonitrile 98%

Supplier: Apollo Scientific

Synonyms: 3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile, 4,5-Dimethoxy-1-cyanobenzocyclobutane, 1-Cyano-4,5-dimethoxybenzocyclobutene

OR929931-10G OR929931-1G OR929931-5G
APOSOR929931-10GEA 720 EUR
APOSOR929931-10G APOSOR929931-1G APOSOR929931-5G
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile 98%
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile
Formula: C₁₁H₁₁NO₂
MW: 189,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01846132
CAS Number: 35202-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR