5-Bromo-1,2,4-thiadiazole 97%
Supplier: Apollo Scientific
|
Warning
|
OR930049-1G
OR930049-5G
APOSOR930049-1GEA
132
EUR
APOSOR930049-1G
APOSOR930049-5G
5-Bromo-1,2,4-thiadiazole 97%
5-Bromo-1,2,4-thiadiazole
Formula:
C₂HBrN₂S MW: 165,0117 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687255 CAS Number: 43201-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...