4,5-Dichloro-2-(methylsulphanyl)pyrimidine 95%

Supplier: Apollo Scientific

Synonyms: 4,5-Dichloro-2-(methylthio)pyrimidine

OR930094-1G OR930094-250MG
APOSOR930094-1GEA 98.9 EUR
APOSOR930094-1G APOSOR930094-250MG
4,5-Dichloro-2-(methylsulphanyl)pyrimidine 95%
4,5-Dichloro-2-(methylsulphanyl)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987934
CAS Number: 99469-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR